Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TACATGCCTATTTATTTGTCACTAA[C/T]AACGACAAAGACCGAGAAGGGCATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 615283 MIM: 616086 | ||||||||||||||||||||
Literature Links: |
EXOC8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EXOC8 - exocyst complex component 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPRTN - SprT-like N-terminal domain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010984.3 | 814 | Silent Mutation | AAC,AAT | N,N 122 | NP_001010984.1 | |
NM_001261462.2 | 814 | Intron | NP_001248391.1 | |||
NM_032018.6 | 814 | Silent Mutation | AAC,AAT | N,N 122 | NP_114407.3 | |
XM_006711818.2 | 814 | Intron | XP_006711881.1 | |||
XM_011544289.2 | 814 | Intron | XP_011542591.1 |