Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTACATCTGCATGGGCTTGCAGTTC[A/G]TACTCTGGTATAGGGGTTTGCCAAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609109 | ||||||||||||||||||||
Literature Links: |
FBXO42 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FBXO42 - F-box protein 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018994.2 | 2170 | Missense Mutation | ACG,ATG | T,M 639 | NP_061867.1 | |
XM_006710698.3 | 2170 | Missense Mutation | ACG,ATG | T,M 639 | XP_006710761.1 | |
XM_017001502.1 | 2170 | Missense Mutation | ACG,ATG | T,M 408 | XP_016856991.1 | |
XM_017001503.1 | 2170 | Missense Mutation | ACG,ATG | T,M 408 | XP_016856992.1 |