Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGACTGCAGTCGCAGGAAATTG[C/T]CCGCACCGAGCTGGAGGGCGCTGTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608869 | ||||||||||||||||||||
Literature Links: |
LOC100996251 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100996251 - uncharacterized LOC100996251 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRIG2 - leucine rich repeats and immunoglobulin like domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001312686.1 | 421 | UTR 5 | NP_001299615.1 | |||
NM_014813.2 | 421 | Missense Mutation | CCC,TCC | P,S 62 | NP_055628.1 | |
XM_005271369.2 | 421 | Missense Mutation | CCC,TCC | P,S 62 | XP_005271426.1 | |
XM_017002952.1 | 421 | Intron | XP_016858441.1 |