Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCAGGATCCTCTCAGCATTCATC[C/T]GGTAGCTGTCTGCAGCCTCGGCCAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604704 MIM: 608848 | ||||||||||||||||||||
Literature Links: |
BCAR3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
BCAR3 - breast cancer anti-estrogen resistance 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001261408.1 | 2164 | Missense Mutation | CAG,CGG | Q,R 758 | NP_001248337.1 | |
NM_001261409.1 | 2164 | Missense Mutation | CAG,CGG | Q,R 758 | NP_001248338.1 | |
NM_001261410.1 | 2164 | Missense Mutation | CAG,CGG | Q,R 667 | NP_001248339.1 | |
NM_001308251.1 | 2164 | Missense Mutation | CAG,CGG | Q,R 434 | NP_001295180.1 | |
NM_003567.3 | 2164 | Missense Mutation | CAG,CGG | Q,R 758 | NP_003558.1 | |
XM_011542251.2 | 2164 | Missense Mutation | CAG,CGG | Q,R 538 | XP_011540553.1 | |
XM_017002480.1 | 2164 | Missense Mutation | CAG,CGG | Q,R 758 | XP_016857969.1 |
FNBP1L - formin binding protein 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001024948.2 | 2164 | Intron | NP_001020119.1 | |||
NM_001164473.2 | 2164 | Intron | NP_001157945.1 | |||
NM_017737.4 | 2164 | Intron | NP_060207.2 | |||
XM_011541625.2 | 2164 | Intron | XP_011539927.1 | |||
XM_017001533.1 | 2164 | Intron | XP_016857022.1 |