Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCTCTCATCTCATCTATTCATTC[A/G]CCAGCATCGAAAACAACAAGGTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601526 | ||||||||||||||||||||
Literature Links: |
CHI3L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHI3L2 - chitinase 3 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025197.1 | 261 | Missense Mutation | ACC,GCC | T,A 54 | NP_001020368.1 | |
NM_001025199.1 | 261 | UTR 5 | NP_001020370.1 | |||
NM_004000.2 | 261 | Missense Mutation | ACC,GCC | T,A 64 | NP_003991.2 |