Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTGACCCCTTCCTATGCTCTCAT[C/T]TCATCTATTCATTCGCCAGCATCGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601526 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHI3L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHI3L2 - chitinase 3 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001025197.1 | 246 | Missense Mutation | CTC,TTC | L,F 49 | NP_001020368.1 | |
NM_001025199.1 | 246 | UTR 5 | NP_001020370.1 | |||
NM_004000.2 | 246 | Missense Mutation | CTC,TTC | L,F 59 | NP_003991.2 |