Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGGTATGTCACTCCACCCTGTGT[C/T]AGCGTCTGTGGGGGCAGCATCCATT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602790 MIM: 605596 | ||||||||||||||||||||
Literature Links: |
FHL3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FHL3 - four and a half LIM domains 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243878.1 | 110 | Silent Mutation | CTA,CTG | L,L 61 | NP_001230807.1 | |
NM_004468.4 | 110 | Silent Mutation | CTA,CTG | L,L 169 | NP_004459.2 | |
XM_017000675.1 | 110 | Silent Mutation | CTA,CTG | L,L 36 | XP_016856164.1 |
SF3A3 - splicing factor 3a subunit 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |