Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGGAGCCAGAGCCAGCTACTCAACA[C/T]CCTAACTAAAAAGCAGAAGAAACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DIEXF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DIEXF - digestive organ expansion factor homolog (zebrafish) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014388.6 | 141 | Missense Mutation | ACC,ATC | T,I 15 | NP_055203.4 | |
XM_006711275.3 | 141 | Missense Mutation | ACC,ATC | T,I 15 | XP_006711338.1 | |
XM_011509418.2 | 141 | UTR 5 | XP_011507720.1 |