Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAAAATGGAGATGTCAGTTGATTT[C/T]AGAAGGTATACCTACAGACAGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605199 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SFPQ PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SFPQ - splicing factor proline and glutamine rich | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005066.2 | Intron | NP_005057.1 | ||||
XM_005271112.4 | Intron | XP_005271169.1 | ||||
XM_005271113.4 | Intron | XP_005271170.1 | ||||
XM_005271115.4 | Intron | XP_005271172.1 | ||||
XM_011541950.2 | Intron | XP_011540252.1 | ||||
XM_017002053.1 | Intron | XP_016857542.1 | ||||
XM_017002054.1 | Intron | XP_016857543.1 |