Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCAGAAAAAATGCTCAATTTTCA[C/T]TGCCAACGTTCTTGGTAGATTCACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNHIT6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNHIT6 - zinc finger HIT-type containing 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170670.1 | 1552 | Missense Mutation | AAT,AGT | N,S 429 | NP_001164141.1 | |
NM_017953.3 | 1552 | Missense Mutation | AAT,AGT | N,S 468 | NP_060423.3 | |
XM_011541614.2 | 1552 | Intron | XP_011539916.1 |