Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCTCTGAGAAGGTTTTCCCTTTC[C/G]TCCTCCAACTTCTTCTCCATTTGGG
Species: |
Human | ||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612466 | ||||||||||||||||||||||||||||||||
Literature Links: |
GBP4 PubMed Links | ||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
GBP4 - guanylate binding protein 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052941.4 | 1769 | Missense Mutation | GAC,GAG | D,E 551 | NP_443173.2 |