Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGGGTACTAATGCGAATGATGCC[A/G]CGTACCAGGCGGCCCTCAGCATCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616758 MIM: 610325 | ||||||||||||||||||||
Literature Links: |
KDF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KDF1 - keratinocyte differentiation factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152365.2 | 990 | Silent Mutation | CGC,CGT | R,R 299 | NP_689578.2 | |
XM_005245735.2 | 990 | Silent Mutation | CGC,CGT | R,R 299 | XP_005245792.1 | |
XM_011540622.1 | 990 | Silent Mutation | CGC,CGT | R,R 299 | XP_011538924.1 |
NUDC - nuclear distribution C, dynein complex regulator | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |