Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTGGCGTGCCATTGTGTGTCAG[C/T]GCTTTCAGACTGGAGGCACTAGTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608267 | ||||||||||||||||||||
Literature Links: |
RRAGC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRAGC - Ras related GTP binding C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271851.1 | 1239 | Silent Mutation | GCA,GCG | A,A 354 | NP_001258780.1 | |
NM_022157.3 | 1239 | Silent Mutation | GCA,GCG | A,A 388 | NP_071440.1 |