Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTCCCTCTAGGGCACCTCTATAAC[A/G]ATCTAGGCCTGGGCTACTCCCAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608862 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IQGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IQGAP3 - IQ motif containing GTPase activating protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAXE - NAD(P)HX epimerase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTC24 - tetratricopeptide repeat domain 24 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105669.2 | 757 | Missense Mutation | AAT,GAT | N,D 241 | NP_001099139.2 | |
XM_011509253.2 | 757 | Missense Mutation | AAT,GAT | N,D 38 | XP_011507555.1 | |
XM_011509254.2 | 757 | Missense Mutation | AAT,GAT | N,D 38 | XP_011507556.1 | |
XM_011509255.2 | 757 | Missense Mutation | AAT,GAT | N,D 38 | XP_011507557.1 | |
XM_017000497.1 | 757 | Intron | XP_016855986.1 |