Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCGACCTCGAAGTCCAGATCCTGA[A/G]TGTCTGCAGCTTCACGGGGAGGCTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 162030 | ||||||||||||||||||||
Literature Links: |
NGF PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NGF - nerve growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002506.2 | 468 | Missense Mutation | ACT,ATT | T,I 100 | NP_002497.2 | |
XM_006710663.3 | 468 | Missense Mutation | ACT,ATT | T,I 100 | XP_006710726.1 | |
XM_011541518.2 | 468 | Missense Mutation | ACT,ATT | T,I 155 | XP_011539820.1 |