Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGATTTCCTACCTGAAGTCGTAG[C/T]TGAGCCATGTCTCTCATGAGCCTGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614579 MIM: 610962 | ||||||||||||||||||||
Literature Links: |
PAQR6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PAQR6 - progestin and adipoQ receptor family member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SMG5 - SMG5, nonsense mediated mRNA decay factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323614.1 | 2585 | Silent Mutation | CAA,CAG | Q,Q 777 | NP_001310543.1 | |
NM_001323615.1 | 2585 | Silent Mutation | CAA,CAG | Q,Q 784 | NP_001310544.1 | |
NM_001323616.1 | 2585 | Silent Mutation | CAA,CAG | Q,Q 764 | NP_001310545.1 | |
NM_001323617.1 | 2585 | Silent Mutation | CAA,CAG | Q,Q 764 | NP_001310546.1 | |
NM_015327.2 | 2585 | Silent Mutation | CAA,CAG | Q,Q 830 | NP_056142.2 | |
XM_017000842.1 | 2585 | Silent Mutation | CAA,CAG | Q,Q 618 | XP_016856331.1 | |
XM_017000843.1 | 2585 | Silent Mutation | CAA,CAG | Q,Q 569 | XP_016856332.1 |