Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCAAGGAGCTAGTCTTGCAAAA[A/G]TTGCACAGAACTTGCTGGGAGACTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616594 MIM: 604128 | ||||||||||||||||||||
Literature Links: |
ASAP3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ASAP3 - ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143778.1 | 2822 | Missense Mutation | CTT,TTT | L,F 891 | NP_001137250.1 | |
NM_017707.3 | 2822 | Missense Mutation | CTT,TTT | L,F 900 | NP_060177.2 | |
XM_011541755.1 | 2822 | Missense Mutation | CTT,TTT | L,F 923 | XP_011540057.1 | |
XM_017001685.1 | 2822 | Missense Mutation | CTT,TTT | L,F 921 | XP_016857174.1 | |
XM_017001686.1 | 2822 | Missense Mutation | CTT,TTT | L,F 902 | XP_016857175.1 | |
XM_017001687.1 | 2822 | Missense Mutation | CTT,TTT | L,F 880 | XP_016857176.1 | |
XM_017001688.1 | 2822 | Missense Mutation | CTT,TTT | L,F 790 | XP_016857177.1 | |
XM_017001689.1 | 2822 | Missense Mutation | CTT,TTT | L,F 771 | XP_016857178.1 |
TCEA3 - transcription elongation factor A3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |