Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCAAATACACAGAGGAAGCCTTC[A/G]CCTGTCCTCATGTATTGGTCTCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 191510 MIM: 164790 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CSDE1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CSDE1 - cold shock domain containing E1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NRAS - neuroblastoma RAS viral oncogene homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002524.4 | 479 | Silent Mutation | GGC,GGT | G,G 75 | NP_002515.1 |