Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGTTCAGTTAGTCACCTTGCTTG[C/T]TGACATTTGAAGATGCATGAATTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611673 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRMT1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRMT1L - tRNA methyltransferase 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001202423.1 | 3460 | Missense Mutation | AAC,AGC | N,S 428 | NP_001189352.1 | |
NM_030934.4 | 3460 | Missense Mutation | AAC,AGC | N,S 584 | NP_112196.3 | |
XM_011510028.2 | 3460 | Missense Mutation | AAC,AGC | N,S 428 | XP_011508330.1 | |
XM_017002419.1 | 3460 | Intron | XP_016857908.1 |