Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGATGCTGTGTGGGCATCACAGCC[C/T]TGGAGGAATAAGGGTCTCCTGGGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 114208 | ||||||||||||||||||||
Literature Links: |
CACNA1S PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNA1S - calcium voltage-gated channel subunit alpha1 S | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000069.2 | 5841 | Missense Mutation | AGG,GGG | R,G 1872 | NP_000060.2 | |
XM_005245478.3 | 5841 | Missense Mutation | AGG,GGG | R,G 1853 | XP_005245535.1 |
LOC101929305 - uncharacterized LOC101929305 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |