Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCAGCTGTCTCCATCCACATTGT[A/G]CAGGCTCTCCCATACGACTTGCTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603319 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANXA9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANXA9 - annexin A9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
FAM63A - family with sequence similarity 63 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040217.2 | 1731 | Missense Mutation | CAC,TAC | H,Y 209 | NP_001035307.1 | |
NM_001163258.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 399 | NP_001156730.1 | |
NM_001163259.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 256 | NP_001156731.1 | |
NM_001163260.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 209 | NP_001156732.1 | |
NM_001319998.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 351 | NP_001306927.1 | |
NM_018379.4 | 1731 | Missense Mutation | CAC,TAC | H,Y 351 | NP_060849.2 | |
XM_005245321.3 | 1731 | Missense Mutation | CAC,TAC | H,Y 352 | XP_005245378.1 | |
XM_005245325.3 | 1731 | Missense Mutation | CAC,TAC | H,Y 210 | XP_005245382.1 | |
XM_005245327.3 | 1731 | Missense Mutation | CAC,TAC | H,Y 186 | XP_005245384.1 | |
XM_005245328.3 | 1731 | Missense Mutation | CAC,TAC | H,Y 185 | XP_005245385.1 | |
XM_011509757.2 | 1731 | Missense Mutation | CAC,TAC | H,Y 352 | XP_011508059.1 | |
XM_011509759.2 | 1731 | Missense Mutation | CAC,TAC | H,Y 298 | XP_011508061.1 | |
XM_017001770.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 351 | XP_016857259.1 | |
XM_017001771.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 351 | XP_016857260.1 | |
XM_017001772.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 351 | XP_016857261.1 | |
XM_017001773.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 351 | XP_016857262.1 | |
XM_017001774.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 351 | XP_016857263.1 | |
XM_017001775.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 298 | XP_016857264.1 | |
XM_017001776.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 298 | XP_016857265.1 | |
XM_017001777.1 | 1731 | Missense Mutation | CAC,TAC | H,Y 399 | XP_016857266.1 |