Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATTCTCAAAATCGAGAGATCATGA[A/G]ACACCTGAAGGCAATTTGTTTTCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610035 | ||||||||||||||||||||
Literature Links: |
VPS45 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
VPS45 - vacuolar protein sorting 45 homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001279353.1 | 412 | Missense Mutation | AAA,AGA | K,R 28 | NP_001266282.1 | |
NM_001279354.1 | 412 | Missense Mutation | AAA,AGA | K,R 28 | NP_001266283.1 | |
NM_001279355.1 | 412 | Intron | NP_001266284.1 | |||
NM_007259.4 | 412 | Missense Mutation | AAA,AGA | K,R 64 | NP_009190.2 | |
XM_017000164.1 | 412 | Missense Mutation | AAA,AGA | K,R 64 | XP_016855653.1 |