Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGGCAATGAAGATGGCAACACTGA[A/G]GTAACAGATTGGCCGAAACCACACT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZNF281 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZNF281 - zinc finger protein 281 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281293.1 | 3687 | Silent Mutation | ACC,ACT | T,T 725 | NP_001268222.1 | |
NM_001281294.1 | 3687 | Silent Mutation | ACC,ACT | T,T 689 | NP_001268223.1 | |
NM_012482.4 | 3687 | Silent Mutation | ACC,ACT | T,T 725 | NP_036614.1 | |
XM_017000887.1 | 3687 | Silent Mutation | ACC,ACT | T,T 725 | XP_016856376.1 | |
XM_017000888.1 | 3687 | Silent Mutation | ACC,ACT | T,T 725 | XP_016856377.1 |