Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCCTCTTTGTCTTGTAGCTGCGG[A/G]CGCTTACCTGCCCCCCCTGCAGCAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606228 | ||||||||||||||||||||
Literature Links: |
AGO1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AGO1 - argonaute 1, RISC catalytic component | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317122.1 | 220 | Missense Mutation | GAC,GGC | D,G 11 | NP_001304051.1 | |
NM_001317123.1 | 220 | UTR 5 | NP_001304052.1 | |||
NM_012199.4 | 220 | Missense Mutation | GAC,GGC | D,G 11 | NP_036331.1 | |
XM_011541236.2 | 220 | Missense Mutation | GAC,GGC | D,G 11 | XP_011539538.1 | |
XM_017001016.1 | 220 | Missense Mutation | GAC,GGC | D,G 11 | XP_016856505.1 | |
XM_017001017.1 | 220 | Missense Mutation | GAC,GGC | D,G 11 | XP_016856506.1 | |
XM_017001018.1 | 220 | Intron | XP_016856507.1 | |||
XM_017001019.1 | 220 | UTR 5 | XP_016856508.1 |