Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAGGCCCTGAATCTGGCGCAGATG[A/C]AGGAGCAGACGCTGCAGTTGGAGCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 612317 | ||||||||||||||||||||
Literature Links: |
ATAD3B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATAD3B - ATPase family, AAA domain containing 3B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317238.1 | 394 | Intron | NP_001304167.1 | |||
NM_031921.5 | 394 | Missense Mutation | AAG,CAG | K,Q 81 | NP_114127.3 | |
XM_005244806.3 | 394 | Missense Mutation | AAG,CAG | K,Q 81 | XP_005244863.1 | |
XM_011542241.2 | 394 | Missense Mutation | AAG,CAG | K,Q 81 | XP_011540543.1 | |
XM_011542243.2 | 394 | Intron | XP_011540545.1 | |||
XM_011542244.1 | 394 | Missense Mutation | AAG,CAG | K,Q 81 | XP_011540546.1 |
ATAD3C - ATPase family, AAA domain containing 3C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |