Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGTCTAGAAACTTCTGATATTTG[C/G]CAATCTTCTCATCCAGTGCTTTAAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602135 MIM: 609365 | ||||||||||||||||||||
Literature Links: |
DNALI1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DNALI1 - dynein axonemal light intermediate chain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNL2 - G protein nucleolar 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001323623.1 | 1977 | Missense Mutation | GCC,GGC | A,G 673 | NP_001310552.1 | |
NM_001323624.1 | 1977 | Missense Mutation | GCC,GGC | A,G 422 | NP_001310553.1 | |
NM_013285.2 | 1977 | Missense Mutation | GCC,GGC | A,G 605 | NP_037417.1 |