Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTGGGTGAAATGAGAGATTATCT[C/G]TTCTCCCTGGAAATGTCTTCAAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616846 MIM: 609890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EMC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EMC1 - ER membrane protein complex subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271427.1 | 3829 | UTR 3 | NP_001258356.1 | |||
NM_001271428.1 | 3829 | UTR 3 | NP_001258357.1 | |||
NM_001271429.1 | 3829 | UTR 3 | NP_001258358.1 | |||
NM_015047.2 | 3829 | UTR 3 | NP_055862.1 | |||
XM_005245787.1 | 3829 | UTR 3 | XP_005245844.1 | |||
XM_005245788.1 | 3829 | UTR 3 | XP_005245845.1 |
LOC101927895 - uncharacterized LOC101927895 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBR4 - ubiquitin protein ligase E3 component n-recognin 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |