Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAGGAAGGCCTTCGGGGACAT[A/G]GTGCCCGGCTTCAGCGTCTTCACCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 164940 | ||||||||||||||||||||
Literature Links: |
FGR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGR - FGR proto-oncogene, Src family tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042729.1 | 1119 | Silent Mutation | ACC,ACT | T,T 297 | NP_001036194.1 | |
NM_001042747.1 | 1119 | Silent Mutation | ACC,ACT | T,T 297 | NP_001036212.1 | |
NM_005248.2 | 1119 | Silent Mutation | ACC,ACT | T,T 297 | NP_005239.1 | |
XM_006710452.2 | 1119 | Silent Mutation | ACC,ACT | T,T 297 | XP_006710515.1 | |
XM_011541010.1 | 1119 | Silent Mutation | ACC,ACT | T,T 297 | XP_011539312.1 | |
XM_011541011.2 | 1119 | Silent Mutation | ACC,ACT | T,T 228 | XP_011539313.1 | |
XM_011541012.1 | 1119 | Silent Mutation | ACC,ACT | T,T 297 | XP_011539314.1 | |
XM_011541013.2 | 1119 | Silent Mutation | ACC,ACT | T,T 297 | XP_011539315.1 | |
XM_011541014.2 | 1119 | Silent Mutation | ACC,ACT | T,T 93 | XP_011539316.1 | |
XM_017000673.1 | 1119 | Silent Mutation | ACC,ACT | T,T 93 | XP_016856162.1 | |
XM_017000674.1 | 1119 | Silent Mutation | ACC,ACT | T,T 93 | XP_016856163.1 |