Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACAGTGACAGAAAATCCTAGAAT[C/T]CCGGATGTGTTCCTACAGGTAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602651 MIM: 606737 | ||||||||||||||||||||
Literature Links: |
NRDC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NRDC - nardilysin convertase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001101662.1 | 3235 | Silent Mutation | GGA,GGG | G,G 971 | NP_001095132.1 | |
NM_001242361.1 | 3235 | Silent Mutation | GGA,GGG | G,G 907 | NP_001229290.1 | |
NM_002525.2 | 3235 | Silent Mutation | GGA,GGG | G,G 1039 | NP_002516.2 | |
XM_005270903.1 | 3235 | Silent Mutation | GGA,GGG | G,G 734 | XP_005270960.1 | |
XM_011541522.1 | 3235 | Silent Mutation | GGA,GGG | G,G 929 | XP_011539824.1 | |
XM_011541523.1 | 3235 | Silent Mutation | GGA,GGG | G,G 839 | XP_011539825.1 | |
XM_011541525.2 | 3235 | Silent Mutation | GGA,GGG | G,G 734 | XP_011539827.1 | |
XM_017001375.1 | 3235 | Silent Mutation | GGA,GGG | G,G 839 | XP_016856864.1 |
OSBPL9 - oxysterol binding protein like 9 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |