Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCAGGAGTAGCCTGCAGAAGCCA[A/G]GGCCTTTCAGACTGTTCCCCAAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
22 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
HSPB11 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HSPB11 - heat shock protein family B (small) member 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRRC42 - leucine rich repeat containing 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256409.1 | 218 | Missense Mutation | AGG,GGG | R,G 42 | NP_001243338.1 | |
NM_052940.4 | 218 | Missense Mutation | AGG,GGG | R,G 42 | NP_443172.1 | |
XM_006710327.3 | 218 | Missense Mutation | AGG,GGG | R,G 42 | XP_006710390.1 | |
XM_006710328.3 | 218 | Missense Mutation | AGG,GGG | R,G 42 | XP_006710391.1 |