Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGTAGGGTTAAAGGTCTGCTTGTT[C/T]GGCCGGAGACCTGAAGAAAAAGTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607956 MIM: 615463 | ||||||||||||||||||||
Literature Links: |
MED8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MED8 - mediator complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001653.2 | 552 | Silent Mutation | CCA,CCG | P,P 79 | NP_001001653.1 | |
NM_052877.4 | 552 | Silent Mutation | CCA,CCG | P,P 168 | NP_443109.2 | |
NM_201542.4 | 552 | Silent Mutation | CCA,CCG | P,P 168 | NP_963836.2 |
SZT2 - seizure threshold 2 homolog (mouse) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |