Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTAGAGGGATAGCCAGAATGAAAC[C/T]CAGGCAGGAGGGCCTGCAAAAACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607956 MIM: 615463 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MED8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MED8 - mediator complex subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001653.2 | 865 | UTR 3 | NP_001001653.1 | |||
NM_052877.4 | 865 | Missense Mutation | AGT,GGT | S,G 273 | NP_443109.2 | |
NM_201542.4 | 865 | UTR 3 | NP_963836.2 |
SZT2 - seizure threshold 2 homolog (mouse) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |