Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGAAGCTTACCGCCAGCATGCGCC[A/G]TGGCTCCCTGTAGCGGATGTGGATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 103180 MIM: 611859 | ||||||||||||||||||||
Literature Links: |
ARF1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARF1 - ADP ribosylation factor 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C1orf35 - chromosome 1 open reading frame 35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MRPL55 - mitochondrial ribosomal protein L55 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321284.1 | 611 | Silent Mutation | NP_001308213.1 | |||
NM_181441.2 | 611 | Silent Mutation | NP_852106.1 | |||
NM_181454.2 | 611 | Silent Mutation | NP_852119.1 | |||
NM_181455.2 | 611 | Silent Mutation | NP_852120.1 | |||
NM_181456.2 | 611 | Silent Mutation | NP_852121.1 | |||
NM_181462.2 | 611 | Silent Mutation | NP_852127.2 | |||
NM_181463.2 | 611 | Silent Mutation | NP_852128.1 | |||
NM_181464.2 | 611 | Silent Mutation | NP_852129.1 | |||
NM_181465.2 | 611 | Silent Mutation | NP_852130.1 | |||
XM_005273059.3 | 611 | Missense Mutation | XP_005273116.1 | |||
XM_005273061.4 | 611 | Missense Mutation | XP_005273118.1 | |||
XM_005273062.3 | 611 | Missense Mutation | XP_005273119.1 | |||
XM_005273063.4 | 611 | Missense Mutation | XP_005273120.1 | |||
XM_011544094.2 | 611 | Missense Mutation | XP_011542396.1 | |||
XM_011544095.2 | 611 | Missense Mutation | XP_011542397.1 |