Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATCTTCACCCCACCATCAGCCAA[A/T]GCGTCCAGAGCCATGAATGCCTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 616406 | ||||||||||||||||||||
Literature Links: |
MIR6741 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR6741 - microRNA 6741 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PYCR2 - pyrroline-5-carboxylate reductase family member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271681.1 | 569 | Silent Mutation | GCA,GCT | A,A 113 | NP_001258610.1 | |
NM_013328.3 | 569 | Silent Mutation | GCA,GCT | A,A 187 | NP_037460.2 |