Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTTTAGAGGATTTCTTCTGGGAG[C/G]TCTTCTTCTGAGAGCGGGTTGGGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611930 | ||||||||||||||||||||
Literature Links: |
ISG20L2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ISG20L2 - interferon stimulated exonuclease gene 20 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303095.1 | 590 | Missense Mutation | ACC,AGC | T,S 140 | NP_001290024.1 | |
XM_005245516.4 | 590 | Missense Mutation | ACC,AGC | T,S 140 | XP_005245573.1 | |
XM_005245518.4 | 590 | Intron | XP_005245575.1 | |||
XM_006711560.3 | 590 | Missense Mutation | ACC,AGC | T,S 140 | XP_006711623.1 | |
XM_006711561.3 | 590 | Missense Mutation | ACC,AGC | T,S 140 | XP_006711624.1 |
RRNAD1 - ribosomal RNA adenine dimethylase domain containing 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |