Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTCTGGTCCTGCCCCATCACCAGG[A/G]GAGTCCCGGCACTGCTCAGGCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616775 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AIM1L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AIM1L - absent in melanoma 1-like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZNF683 - zinc finger protein 683 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114759.2 | 1801 | Missense Mutation | CCC,CTC | P,L 495 | NP_001108231.1 | |
NM_001307925.1 | 1801 | Missense Mutation | CCC,CTC | P,L 515 | NP_001294854.1 | |
NM_173574.3 | 1801 | Missense Mutation | CCC,CTC | P,L 495 | NP_775845.2 | |
XM_005245828.3 | 1801 | Missense Mutation | CCC,CTC | P,L 523 | XP_005245885.1 | |
XM_005245830.3 | 1801 | Missense Mutation | CCC,CTC | P,L 515 | XP_005245887.1 | |
XM_005245832.3 | 1801 | Missense Mutation | CCC,CTC | P,L 515 | XP_005245889.1 | |
XM_006710555.3 | 1801 | Missense Mutation | CCC,CTC | P,L 524 | XP_006710618.1 | |
XM_011541198.2 | 1801 | Missense Mutation | CCC,CTC | P,L 515 | XP_011539500.1 | |
XM_017000954.1 | 1801 | Missense Mutation | CCC,CTC | P,L 524 | XP_016856443.1 | |
XM_017000955.1 | 1801 | Missense Mutation | CCC,CTC | P,L 524 | XP_016856444.1 | |
XM_017000956.1 | 1801 | Missense Mutation | CCC,CTC | P,L 523 | XP_016856445.1 | |
XM_017000957.1 | 1801 | Missense Mutation | CCC,CTC | P,L 504 | XP_016856446.1 | |
XM_017000958.1 | 1801 | UTR 3 | XP_016856447.1 | |||
XM_017000959.1 | 1801 | UTR 3 | XP_016856448.1 |