Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTGTCAGACCTAGAGTTGTTATTT[C/T]GTTTCTACCTCTTTGGATTAACTCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 188360 MIM: 188340 | ||||||||||||||||||||
Literature Links: |
CD1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD1B - CD1b molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001764.2 | 557 | Intron | NP_001755.1 | |||
XM_011510119.2 | 557 | Intron | XP_011508421.1 | |||
XM_017002784.1 | 557 | Intron | XP_016858273.1 | |||
XM_017002785.1 | 557 | Intron | XP_016858274.1 | |||
XM_017002786.1 | 557 | Intron | XP_016858275.1 |
CD1C - CD1c molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001765.2 | 557 | Missense Mutation | CGT,TGT | R,C 89 | NP_001756.2 | |
XM_005245579.4 | 557 | Missense Mutation | CGT,TGT | R,C 89 | XP_005245636.1 |