Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAAAAGCTCAGGTGCTTGGAAGTTC[C/T]CCTGGGGTCCCTTGGGATTCCTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GPR157 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GPR157 - G protein-coupled receptor 157 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024980.4 | 887 | Silent Mutation | GGA,GGG | G,G 330 | NP_079256.4 | |
XM_005263496.4 | 887 | Silent Mutation | GGA,GGG | G,G 317 | XP_005263553.1 | |
XM_005263497.4 | 887 | Silent Mutation | GGA,GGG | G,G 265 | XP_005263554.1 |