Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACCTTGCTGCGGAGAAGCCCCCC[A/G]CCCTGGTGCTCTGGTGTGCTGGTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611906 MIM: 611889 | ||||||||||||||||||||
Literature Links: |
FNDC5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FNDC5 - fibronectin type III domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001171940.1 | 679 | Intron | NP_001165411.2 | |||
NM_001171941.2 | 679 | Silent Mutation | GGC,GGT | G,G 129 | NP_001165412.1 | |
NM_153756.2 | 679 | Silent Mutation | GGC,GGT | G,G 204 | NP_715637.2 |
S100PBP - S100P binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |