Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602105 MIM: 179080 | ||||||||||||||||||||
Literature Links: |
MSH4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MSH4 - mutS homolog 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002440.3 | 128 | Missense Mutation | CAG,GAG | Q,E 5 | NP_002431.2 |
RABGGTB - Rab geranylgeranyltransferase beta subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD45A - small nucleolar RNA, C/D box 45A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD45B - small nucleolar RNA, C/D box 45B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD45C - small nucleolar RNA, C/D box 45C | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |