Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCCTGCTCACCAGAATCTATACC[A/G]AGAGGTGATGCTGGAGAACTACAGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ZFP69 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ZFP69 - ZFP69 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320178.1 | 1306 | Missense Mutation | CAA,CGA | Q,R 102 | NP_001307107.1 | |
NM_001320179.1 | 1306 | Missense Mutation | CAA,CGA | Q,R 102 | NP_001307108.1 | |
NM_198494.2 | 1306 | Missense Mutation | CAA,CGA | Q,R 102 | NP_940896.2 | |
XM_005270809.4 | 1306 | Missense Mutation | CAA,CGA | Q,R 102 | XP_005270866.1 | |
XM_006710605.3 | 1306 | Missense Mutation | CAA,CGA | Q,R 102 | XP_006710668.1 | |
XM_006710606.3 | 1306 | Missense Mutation | CAA,CGA | Q,R 102 | XP_006710669.1 |