Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCCATTGGTTTTGTCGCTGTCGT[C/T]AACTGACAGTGATTCATCACTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603538 MIM: 606695 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KMO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KMO - kynurenine 3-monooxygenase (kynurenine 3-hydroxylase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003679.4 | 1295 | UTR 3 | NP_003670.2 | |||
XM_005273337.1 | 1295 | Intron | XP_005273394.1 | |||
XM_005273338.3 | 1295 | Intron | XP_005273395.1 | |||
XM_017002649.1 | 1295 | Intron | XP_016858138.1 | |||
XM_017002650.1 | 1295 | Intron | XP_016858139.1 |
OPN3 - opsin 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014322.2 | 1295 | Missense Mutation | AAC,GAC | N,D 384 | NP_055137.2 |