Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTACGGCCTTGAGCTGTCTCCTAG[A/G]CCTCCCCTTGCGGGTGCAGAAGATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611286 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RTCA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RTCA - RNA 3'-terminal phosphate cyclase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130841.1 | 229 | Missense Mutation | GAC,GGC | D,G 31 | NP_001124313.1 | |
NM_003729.3 | 229 | Missense Mutation | GAC,GGC | D,G 31 | NP_003720.1 | |
XM_005271298.4 | 229 | UTR 5 | XP_005271355.1 | |||
XM_005271299.4 | 229 | UTR 5 | XP_005271356.1 | |||
XM_017002669.1 | 229 | Intron | XP_016858158.1 |
RTCA-AS1 - RTCA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |