Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATATTTCAGGATCTTTTTCTTTGGG[A/G]CTATCGTTGCCTGGGAAAGGCTGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600051 | ||||||||||||||||||||
Literature Links: |
EPS15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EPS15 - epidermal growth factor receptor pathway substrate 15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001159969.1 | 2475 | Silent Mutation | AGC,AGT | S,S 500 | NP_001153441.1 | |
NM_001981.2 | 2475 | Silent Mutation | AGC,AGT | S,S 814 | NP_001972.1 | |
XM_005270618.2 | 2475 | Silent Mutation | AGC,AGT | S,S 784 | XP_005270675.1 | |
XM_017000615.1 | 2475 | Silent Mutation | AGC,AGT | S,S 851 | XP_016856104.1 | |
XM_017000616.1 | 2475 | Silent Mutation | AGC,AGT | S,S 783 | XP_016856105.1 | |
XM_017000617.1 | 2475 | Silent Mutation | AGC,AGT | S,S 746 | XP_016856106.1 | |
XM_017000618.1 | 2475 | Silent Mutation | AGC,AGT | S,S 716 | XP_016856107.1 | |
XM_017000619.1 | 2475 | Silent Mutation | AGC,AGT | S,S 626 | XP_016856108.1 | |
XM_017000620.1 | 2475 | Silent Mutation | AGC,AGT | S,S 576 | XP_016856109.1 |