Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTTTCTGCCTTCTCTAGGCTG[A/C]GAACACAATCACCTATTCACTTCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604590 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FCGR2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FCGR2B - Fc fragment of IgG receptor IIb | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001002273.2 | 987 | Missense Mutation | GAG,GCG | E,A 267 | NP_001002273.1 | |
NM_001002274.2 | 987 | Missense Mutation | GAG,GCG | E,A 268 | NP_001002274.1 | |
NM_001002275.2 | 987 | Missense Mutation | GAG,GCG | E,A 286 | NP_001002275.1 | |
NM_001190828.1 | 987 | Missense Mutation | GAG,GCG | E,A 280 | NP_001177757.1 | |
NM_004001.4 | 987 | Missense Mutation | GAG,GCG | E,A 287 | NP_003992.3 | |
XM_011509292.2 | 987 | Intron | XP_011507594.1 | |||
XM_017000669.1 | 987 | Intron | XP_016856158.1 | |||
XM_017000670.1 | 987 | Intron | XP_016856159.1 | |||
XM_017000671.1 | 987 | Missense Mutation | GAG,GCG | E,A 287 | XP_016856160.1 | |
XM_017000672.1 | 987 | Missense Mutation | GAG,GCG | E,A 268 | XP_016856161.1 |
RPL31P11 - ribosomal protein L31 pseudogene 11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |