Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGAACTCTCGTCTGCTCATTGAAG[G/T]ACAAGATGAAGACGCAGGTGGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601883 | ||||||||||||||||||||
Literature Links: |
C1orf174 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf174 - chromosome 1 open reading frame 174 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_207356.2 | 3241 | Missense Mutation | ACT,CCT | T,P 216 | NP_997239.2 | |
XM_005244743.3 | 3241 | Missense Mutation | ACT,CCT | T,P 227 | XP_005244800.1 | |
XM_011541323.2 | 3241 | Missense Mutation | ACT,CCT | T,P 227 | XP_011539625.1 |
DFFB - DNA fragmentation factor subunit beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |