Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCCGTCGCGGAGCCAGATCACCTG[A/T]GGGAAGCGGGCATCACGGCCGTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604835 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DUSP12 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DUSP12 - dual specificity phosphatase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007240.2 | 179 | Missense Mutation | AGG,TGG | R,W 49 | NP_009171.1 | |
XM_005244862.2 | 179 | UTR 5 | XP_005244919.1 |