Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCGGGGAATGTGACATCAGCGGC[G/T]CCGGGCGCTTGGGGCTGGAGGAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607813 | ||||||||||||||||||||
Literature Links: |
PLPPR4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PLPPR4 - phospholipid phosphatase related 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166252.1 | 210 | Missense Mutation | GCC,TCC | A,S 18 | NP_001159724.1 | |
NM_014839.4 | 210 | Missense Mutation | GCC,TCC | A,S 18 | NP_055654.2 | |
XM_011542498.2 | 210 | Intron | XP_011540800.1 |