Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGACCCTCACTCTAGGGGACAAG[A/G]CCTCTATCCCAGGGAACTCCACCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
EVA1B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
EVA1B - eva-1 homolog B (C. elegans) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3D21 - SH3 domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162530.1 | 1259 | Missense Mutation | ACC,GCC | T,A 396 | NP_001156002.1 | |
NM_024676.4 | 1259 | Missense Mutation | ACC,GCC | T,A 285 | NP_078952.4 | |
XM_011542153.2 | 1259 | Missense Mutation | ACC,GCC | T,A 396 | XP_011540455.1 | |
XM_017002340.1 | 1259 | Missense Mutation | ACC,GCC | T,A 396 | XP_016857829.1 | |
XM_017002341.1 | 1259 | Missense Mutation | ACC,GCC | T,A 396 | XP_016857830.1 | |
XM_017002342.1 | 1259 | Missense Mutation | ACC,GCC | T,A 396 | XP_016857831.1 | |
XM_017002343.1 | 1259 | Missense Mutation | ACC,GCC | T,A 396 | XP_016857832.1 | |
XM_017002344.1 | 1259 | Missense Mutation | ACC,GCC | T,A 396 | XP_016857833.1 | |
XM_017002345.1 | 1259 | Missense Mutation | ACC,GCC | T,A 396 | XP_016857834.1 | |
XM_017002346.1 | 1259 | Missense Mutation | ACC,GCC | T,A 42 | XP_016857835.1 |