Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTGAGAACGTCATCTGGATTGGTC[C/T]CAAAAATGCTCTTGCTTTCGCACGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600684 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105371470 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105371470 - uncharacterized LOC105371470 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LY9 - lymphocyte antigen 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001033667.2 | 362 | Missense Mutation | CCC,CTC | P,L 83 | NP_001028839.1 | |
NM_001261456.1 | 362 | Missense Mutation | CCC,CTC | P,L 83 | NP_001248385.1 | |
NM_001261457.1 | 362 | Missense Mutation | CCC,CTC | P,L 83 | NP_001248386.1 | |
NM_002348.3 | 362 | Missense Mutation | CCC,CTC | P,L 83 | NP_002339.2 | |
XM_011509548.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_011507850.1 | |
XM_011509549.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_011507851.1 | |
XM_011509550.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_011507852.1 | |
XM_011509552.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_011507854.1 | |
XM_011509556.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_011507858.1 | |
XM_011509560.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_011507862.1 | |
XM_017001297.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_016856786.1 | |
XM_017001298.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_016856787.1 | |
XM_017001299.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_016856788.1 | |
XM_017001300.1 | 362 | Missense Mutation | CCC,CTC | P,L 97 | XP_016856789.1 | |
XM_017001301.1 | 362 | Missense Mutation | CCC,CTC | P,L 83 | XP_016856790.1 | |
XM_017001302.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_016856791.1 | |
XM_017001303.1 | 362 | Missense Mutation | CCC,CTC | P,L 130 | XP_016856792.1 | |
XM_017001304.1 | 362 | UTR 5 | XP_016856793.1 |